missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Labeled Wash Bottles
Click to view available options
Capacity (Metric):
500 mL
Description
- Easy-to-read printed bottles
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Translucent, LDPE solvent-resistant
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
- Toluene is red LDPE bottle
- Polypropylene closure with leakproof design
- Bottles can be purchased separately or in variety packs that include water, acetone, methanol, isopropanol, toluene and ethanol
Specifications
Capacity | 500mL (16oz) |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Diameter (English) Outer Mouth | 2 in. |
Diameter (Metric) Outer Mouth | 53mm |
Material | LDPE |
Quantity | 6 |
Type | Wash Bottle |
Certifications | Hazard Communication Standard 29 CFR 1910.1200 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'