missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Labmaster Air-Drive Mixer

Product Code. 15467463 Shop All Cole-Parmer Products
Click to view available options
Mounting Type:
Tube Suspension
Universal Clamp
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15467463

Brand: Cole-Parmer™ 5032210

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use Labmaster Air-Drive Mixers in the presence of volatile or explosive vapors, or where electrical power would be hazardous or impractical. These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes.

  • These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes.
  • LABMASTER air-drive mixers have two types of mounting styles.
    • The tube suspension mixers require the heavy-duty clamp 04560-24 (sold separately).
    • The universal clamp mixers mount directly to a ring stand or tank rim.
  • Mixers include:
    • 5/16 x 12 in., 316 SS nonthrough-type shaft
    • 5/16 x 2.5 in., 316 SS A-310 impeller
    • Collet-type chuck
    • Universal clamp or tube suspension mounting
    • Muffler
    • Needle value
  • Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Specifications

Speed 1800 rpm
Horsepower 1/15 hp
Material 316 Stainless Steel
Motor Type Air-Driven
Torque 37 in. oz.
Type Mixer
Mounting Type Universal Clamp
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.