Learn More
SHP Steriltechnik AG Laboklav 160 Trolley
Product Code. 31908806
Change view
Click to view available options
Capacity (Metric):
300 kg
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
31908806 300 kg Each
1 options
Product Code. 31908806 Supplier SHP Steriltechnik AG Supplier No. 20.0160.000.301
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Trolley for Laboklav 160, horizontal unit.
  • Stainless Steel
  • With 4 castors for easy transport
Specifications
Product Type Trolley for Steam Sterilizer
Capacity (Metric) 300 kg
For Use With (Equipment) Laboklav 160