Learn More
Hirschmann™ Labopette™ Variable Volume Single Channel Manual Pipette
Product Code. 10831271
Click to view available options
Volume (Metric):
10 to 100 μL
100 to 1000 μL
20 to 200 μL
500 to 5000 μL
Unit Size:
Each
Product Code. 10831271
Brand: Hirschmann™ 9475405
Only null left
Add to basket
Add to basket
Ergonomically shaped manual pipette, effortless operation and tip ejector.
  • Variable volume setting
  • With tip ejector and recalibration possibility
  • Autoclavable
Specifications
Accuracy 0.8%
Certifications/Compliance CE, IVD 98/79 EC
Compatible Tips 100 to 5000 μL
Autoclavable Autoclavable
Color Clear
Number of Channels 1
Product Line Labopette
Type Labopettor
Volume (Metric) 500 to 5000 μL
Includes Calibration tool and pipette holder, individual serial number and calibration certificate
Increments 10 μL