Learn More
Bochem™ Laboral Squared Bossheads
Product Code. 10646951
Click to view available options
Includes:
Allen screws
Shape:
Square
Size:
12 to 13 mm
Product Type:
Laboral Bossheads
Unit Size:
Each
Product Code. 10646951
Brand: Bochem™ B08600121
Only null left
Add to basket
Edge
Add to basket
Keep clamps in place. BOCHEM™ Laboral Squared Bossheads come with hexagon socket screws.
  • With hexagon socket screws
  • Combination possibility for adjustment of the angle from 0-360°
Specifications
Includes Allen screws
Product Type Laboral Bossheads
Shape Square
Size 12 to 13 mm
Quantity 1 Ea.