Learn More
Buerkle™ Laboratory Bottle PET Sterile
Product Code. p-7235925 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Unit Size:
Each
Product Code. 15722785
Brand: Buerkle™ 03700125
Only null left
Add to basket
Edge
Add to basket
Sterile laboratory bottle for use in laboratories and chemical handling areas. Good product presentation due to crystal-clear PET. Space-saving and absolutely leakproof. Ideal for storing and packing liquids.
  • Sterile SAL 10-3 (according to ISO 11137)
  • PET crystal-clear
  • With tamper-proof closure (HDPE)
  • With moulded-in raised graduation
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 125 mL
Material PET