missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments Laboratory Centrifuge

Product Code. 30274337
Change view
Click to view available options
Capacity:
8 x 50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
30274337 8 x 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30274337 Supplier Grant Instruments Supplier No. LMC56 EU
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Modern benchtop, low-speed laboratory centrifuge designed for convenient sedimentation, centrifugation and collection of samples

Our centrifuges are designed for safe work, easy maintenance and wide application range in medical, biochemical, chemical, industrial and other type of laboratories.

Features:

  • User-friendly displays shows set and actual values.
  • Safe assay performance including metal protective housing and metal lid, automatic imbalance switch-off and lid lock during the centrifuge operation provide safe operation at all speeds.
  • Rotor imbalance automatic diagnostics (emergency stop, imbalance indication).
  • Automatic rotor detection with active rotational speed limit.
  • Maximum speed of 6,000 rpm.
  • Wide choice of accessory rotors and adapters.
  • Improved chamber to reduce sample heating during centrifugation.
  • Different modes of acceleration and deceleration, including deceleration mode with switched off forced braking.
  • Possibility to set the speed both in revolutions per minute and by relative centrifugal force.
  • It provides operation with tubes, blood collection systems, gel cards, microtest and ELISA plates.
TRUSTED_SUSTAINABILITY

Specifications

Acceleration Time 280 rpm/sec.
Certifications/Compliance CE
Controller Type Mounted Membrane Controller
Capacity 8 x 50 mL
Max. RCF 3,750 x g
Max. Speed 6,000 rpm
Refrigerated No
Drive System Brushless, Direct
Includes LMC-56
Material Stainless steel
Model LMC-56
Noise Level 65 dB
Quantity 1 Ea.
Rotor Included No
Run Time 1 to 90 min. or Non-stop
Timer 1 to 90 min.
Warranty 2 Years
Dimensions (L x W x H) Exterior 480 x 560 x 315 mm
Length (English) Exterior 19 in.
Length (Metric) Exterior 480 mm
Width (English) Exterior 22 in.
Width (Metric) Exterior 560 mm
Height (English) Exterior 12.4 in.
Height (English) Exterior Lid Open 23.6 in.
Height (Metric) Exterior 315 mm
Height (Metric) Exterior Lid Open 600 mm
Depth (English) 19 in.
Depth (Metric) 480 mm
Net Weight 16.5 kg
Voltage 230 V
Wattage 320 W
Amperage 1.39 A
Frequency 50/60 Hz
Laboratory/Cleanroom Use Yes
Plug Type EU
Product Type Universal Centrifuge
Display Type LCD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.