missing translation for 'onlineSavingsMsg'
Learn More

Heinz Herenz Laboratory Dish Set 4 Pieces

Product Code. 30158302
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
38 mm
missing translation for 'unitSize'
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30158302 38 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30158302 missing translation for 'mfr' Heinz Herenz missing translation for 'supplierNo' 1083295
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory dish set.

  • Laboratory dish set
  • 4 piece.set: 0.5 L - 180 x 230 x 42 mm, 1.5 L - 250 x 310 x 65 mm, 3.0 L - 310 x 370 x 75 mm, 10.0 L - 420 x 520 x 120 mm
  • Plastic

Specifications

Autoclavable Autoclavable
Bottom Shape Flat
Capacity (Metric) 38 mm
Color White
Disposable Reusable
Shape Rectangular
Sterility Non-sterile
Surface Treatment Non-treated
Type Laboratory Dish
Height (Metric) 120 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.