Learn More
Miele Professional™ Lab Washer PG 8583 WW APD LD
Optimally designed laboratory washer with perfectly aligned wash pressure and ideally designed baskets, filters, and spray arms. Miele™ PG 8583 WW APD LD Laboratory Glassware Washer features a variety of specific wash programs to meet multiple laboratory needs.
Brand: Miele Professional™ 62858326
This item is not returnable.
View return policy
Description
- Variable-speed heater pump
- EcoDry via patented AutoOpen function
- Optimum cleaning performance
- Optimum wash pressure
- Low consumption values
- Short batch times
- Crevice-free wash cabinet
- Spray arm pressure monitoring
- Spray arm monitoring
- Hot water connection
- De-min. water connection
- Liquid dispensing
- Conductivity monitoring