missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Laboratory Manifold Standard Adapter for Number 8 Stopper
Description
- Lab manifold is designed for flexibility and reduction in cross-contamination
- Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | Number 8 Stopper |
| Material | Fluoroelastomer |
| Sterility | Non-sterile |
| No. per Pack | 3/Pk. |
| Type | Standard Adapter |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?