missing translation for 'onlineSavingsMsg'
Learn More

Melford Lactic Acid 88% Solution

Product Code. 30018369
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018369 1 L 1 L
30018371 500 mL 500mL
30018370 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018369 Supplier Melford Supplier No. L175001000.0
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ACS Reagent Grade

ACS Reagent Grade

Specifications

Chemical Name or Material Lactic Acid
CAS 50-21-5
Purity Grade Notes ACS Reagent Grade
Molecular Formula C3H6O3
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.