missing translation for 'onlineSavingsMsg'
Learn More

Lactic acid, Solstice
BIOPHARMA

Product Code. 15636730
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15636730 250 mL 250mL
15626730 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15636730 Supplier Honeywell-Fluka Supplier No. 69775250ML
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tested according to Ph.Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Density 1.209g/mL at 25°C
Quantity 250 mL
Flash Point 113°C (235.4°F)
Linear Formula CH3CH(OH)COOH
UN Number NONH for all modes of transport
Beilstein 1209341
Formula Weight 90.08g/mol
Chemical Name or Material Lactic acid
compliance-icons
Product Identifier
  • Lactic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Lactic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.