missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Lactophenol Cotton Blue Stain, 500mL

Product Code. 12811823
Click to view available options
Quantity:
500mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12811823

Brand: Pro-Lab Diagnostics™ PL.7054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pro-Lab Diagnostics™ Lactophenol Cotton Blue Stain, 500mL is used for preparing wet mounts in the microscopic examination of yeasts and molds, acting as a mounting fluid and stain.

Lactophenol Cotton Blue Stain is formulated with lactophenol, which serves as a mounting fluid, and cotton blue, an acid stain. Organisms suspended in the staining fluid are killed due to the presence of phenol, but the cells do not lyse. Cotton blue stains the cell walls of fungi.

Specifications

Solution Type Reagent
Type Lactophenol Cotton Blue
Quantity 500mL
Certifications/Compliance (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark
Format Liquid
For Use With (Application) For mscellaneous specialty staining (e.g. as a mounting fluid and stain in examining yeast and molds)
Color Blue
Composition Glycerol, Lactic acid
Storage Requirements 2 to 40°C
Packaging Type Bottle
Stain Type Miscellaneous Specialty Stain
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lactophenol Cotton Blue
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash contaminated skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : lactic acid, phenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.