missing translation for 'onlineSavingsMsg'
Learn More

Lactose octaacetate, 95%, BLD Pharm™

Product Code. 31109230
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31109230 100 g
31093206 1 g
31112090 25 g
31093357 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31109230 Supplier BLD Pharm Supplier No. BD135780100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6291-42-5
Molecular Formula C28H38O19
MDL Number MFCD00064040
Quantity 100 g
SMILES CC(OC[C@H]1O[C@H]([C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H]1OC(C)=O)O[C@H]2[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H](OC(C)=O)O[C@@H]2COC(C)=O)=O
Molecular Weight (g/mol) 678.59
Percent Purity 95%
Chemical Name or Material Lactose octaacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.