missing translation for 'onlineSavingsMsg'
Learn More

Lactulose, MP Biomedicals™

Product Code. 11497372 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11497372 100 g 100g
11487372 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11497372

Brand: MP Biomedicals 0215393280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the treatment of chronic constipation, SIBO and hepatic encephalopathy. MP Biomedicals' Lactulose is a synthetic, non-digestible disaccharide (double-sugar) formed from one molecule each of fructose and galactose (monosaccharides). It is also used

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4618-18-2
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
InChI Key JCQLYHFGKNRPGE-DJYZFUSFSA-N
Synonym lactulose
PubChem CID 131839611
IUPAC Name (2S,3S,4S,5R,6R)-2-[(2R,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C2O)(CO)O)CO)O)O)O)O

Specifications

pH 5.5 to 6.5 (1% aq. soln.)
Quantity 100 g
Solubility Information Soluble in 5% of water (colorless)
Optical Rotation −45 to −52° (c=5, water, 24 hrs)
Formula Weight 342.3
Percent Purity ∼98%
Physical Form Powder
compliance-icons compliance-icons
Product Identifier
  • Lactulose
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H370-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Lactulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.