missing translation for 'onlineSavingsMsg'
Learn More

Laduviglusib trihydrochloride, MedChemExpress

Product Code. 16453598
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16453598 10 mg 10mg
16439996 50 mg 50mg
16416327 5 mg 5mg
3 options
This item is not returnable. View return policy

Product Code. 16453598

Brand: Medchem Express HY10182B10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Laduviglusib (CHIR-99021) trihydrochloride is a potent and selective GSK-3α/β inhibitor with IC50s of 10 nM and 6.7 nM.

Laduviglusib trihydrochloride shows >500-fold selectivity for GSK-3 over CDC2, ERK2 and other protein kinases. Laduviglusib trihydrochloride is also a potent Wnt/β-catenin signaling pathway activator. Laduviglusib trihydrochloride enhances mouse and human embryonic stem cells self-renewal. Laduviglusib trihydrochloride induces autophagy.

Specifications

Chemical Name or Material Laduviglusib trihydrochloride
CAS 1782235-14-6
Color White
Purity Grade Notes Research
Molecular Formula C22H21Cl5N8
Quantity 10 mg
Synonym CHIR-99021 trihydrochloride CT99021 trihydrochloride
Solubility Information DMSO : ≥ 32 mg/mL (55.68 mM) ∣H2O : 19 mg/mL (33.06 mM; Need ultrasonic and warming)
SMILES N#CC1=CC=C(NCCNC2=NC=C(C3=NC=C(C)N3)C(C4=CC=C(Cl)C=C4Cl)=N2)N=C1.[H]Cl.[H]Cl.[H]Cl
Molecular Weight (g/mol) 574.72
Formula Weight 574.72
Percent Purity 98.01%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.