missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific Chemicals Laemmli SDS sample buffer, non-reducing (6X)

Artikelnummer. 15413399
Klik for at se tilgængelige muligheder
Quantity:
25 mL
50 mL
Pakningsstørrelse:
25mL
50mL
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 15413399

Brand: Thermo Scientific Chemicals J60660.AC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Laemmli SDS sample buffer is used to prepare protein samples in the Laemmli SDS-PAGE system.

Laemmli SDS sample buffer, non-reducing (6X) is an electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Composition of this buffer is similar to the reducing buffer minus mercaptoethanol. This product is ideal for polyacrylamide protein gel analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Laemmli SDS sample buffer is composed of several compounds, including 375 mM Tris-HCl (pH 6.8), 9% SDS, 50% glycerol, 9% β-mercaptoethanol, 0.03% bromophenol blue
  • The SDS compound present in the Laemmli SDS sample buffer binds noncovalently to proteins. SDS is negatively charged and can mask the intrinsic charge of the protein it binds

Application

  • Laemmli SDS sample buffer is used to denature proteins and for protein sample preparation to be used in the SDS-PAGE system
  • This sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of the running gel
  • It is also used for polyacrylamide protein gel analysis
TRUSTED_SUSTAINABILITY

Tekniske data

Color Blue
Concentration 6X
Product Type Sample Loading Buffer
Form Liquid
For Use With (Application) Laemmli SDS-PAGE
Quantity 25 mL
compliance-icons
Produktidentifikator
  • Laemmli SDS sample buffer, non-reducing (6X)
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 1
Faresætning
  • H318-Causes serious eye damage.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplerende oplysninger
  • MIXTURE LIST-Contains : 1,2,3-Propanetriol

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.