missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lambda Exonuclease (10 U/μL)

Product Code. 10520081 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1,000 units
5,000 units
Unit Size:
1000 units
5000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10520081 1,000 units 1000 units
10344530 5,000 units 5000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10520081 Supplier Thermo Scientific™ Supplier No. EN0561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lambda Exonuclease is a highly processive exodeoxyribonuclease that selectively digests the 5'-phosphorylated strand of double-stranded DNA in 5'-3' direction.

Thermo Scientific Lambda Exonuclease is a highly processive 5'≥3' exodeoxyribonuclease. It selectively digests the 5'-phosphorylated strand of double-stranded DNA. The enzyme exhibits low activity on single-stranded DNA and non-phosphorylated DNA, and has no activity at nicks and limited activity at gaps in DNA.

Highlights

  • Active in Thermo Scientific PCR buffers

Applications

  • Generating single-stranded PCR products for use in:
    • DNA sequencing
    • Analysis of DNA single-strand conformation polymorphism (SSCP)
    • Rolling circle amplification
     
  • Producing single-stranded DNA from double-stranded DNA fragments
  • Cloning of PCR products

Note

Use of this enzyme in certain applications may be covered by patents and may require a license.

TRUSTED_SUSTAINABILITY

Specifikationer

Concentration 10 U/μL
Enzyme Exonuclease
Quantity 1,000 units
Product Type Exonuclease

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.