missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Facility Safety and Maintenance
- Signs and Tags
- Chemical Hazard Signs
- Brady™ Laminated Polyester GHS Symbol Signs
Brady™ Laminated Polyester GHS Symbol Signs
Product Code. 15404851 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
40 mm
50 mm
70 mm
100 mm
Displayed Text:
Acute Toxicity
Compressed Gas
Corrosive
Explosive
Flammable
Hazardous to Aquatic Environment
Health Hazard
Oxidizing
Respiratory Hazard
Style:
Card Mounted
Continuous Roll, 250 Symbols (not labels)/Roll
Individual Markers on a Roll
Quantity:
1 Ea.
100/Pk.
20/Pk.
250/Pk.
6/Pk.
Unit Size:
1 roll
Pack of 20
Pack of 250
Pack of 4
Pack of 6
72 product options available for selection
| Product Code. | Width (Metric) | Displayed Text | Style | Quantity | Unit Size |
|---|---|---|---|---|---|
| 15404851 | 40 mm | Flammable | Card Mounted | 20/Pk. | Pack of 20 |
| 15414871 | 40 mm | Acute Toxicity | Card Mounted | 20/Pk. | Pack of 20 |
| 15404871 | 100 mm | Acute Toxicity | Card Mounted | 20/Pk. | Pack of 4 |
| 11598904 | 100 mm | Acute Toxicity | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15604227 | 100 mm | Acute Toxicity | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 11753889 | 50 mm | Acute Toxicity | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | 1 roll |
| 15614227 | 50 mm | Acute Toxicity | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | Pack of 250 |
| 15434871 | 70 mm | Acute Toxicity | Card Mounted | 6/Pk. | Pack of 6 |
| 15454851 | 100 mm | Compressed Gas | Card Mounted | 20/Pk. | Pack of 4 |
| 11548884 | 100 mm | Compressed Gas | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15604217 | 100 mm | Compressed Gas | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15464851 | 40 mm | Compressed Gas | Card Mounted | 100/Pk. | Pack of 20 |
| 15368751 | 50 mm | Compressed Gas | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15644237 | 50 mm | Compressed Gas | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15474851 | 70 mm | Compressed Gas | Card Mounted | 6/Pk. | Pack of 6 |
| 11763879 | 50 mm | Compressed Gas | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | 1 roll |
| 15614217 | 50 mm | Compressed Gas | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | Pack of 250 |
| 15474861 | 100 mm | Corrosive | Card Mounted | 20/Pk. | Pack of 4 |
| 11548904 | 100 mm | Corrosive | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15684217 | 100 mm | Corrosive | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15484861 | 40 mm | Corrosive | Card Mounted | 20/Pk. | Pack of 20 |
| 11743889 | 50 mm | Corrosive | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | 1 roll |
| 15694217 | 50 mm | Corrosive | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | Pack of 250 |
| 15318761 | 50 mm | Corrosive | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15684237 | 50 mm | Corrosive | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15328751 | 50 mm | Explosive | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15614237 | 50 mm | Explosive | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15494861 | 70 mm | Corrosive | Card Mounted | 6/Pk. | Pack of 6 |
| 15338751 | 50 mm | Flammable | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15624237 | 50 mm | Flammable | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15414861 | 100 mm | Hazardous to Aquatic Environment | Card Mounted | 20/Pk. | Pack of 4 |
| 11548894 | 100 mm | Hazardous to Aquatic Environment | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15644217 | 100 mm | Hazardous to Aquatic Environment | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15424861 | 40 mm | Hazardous to Aquatic Environment | Card Mounted | 20/Pk. | Pack of 20 |
| 11723889 | 50 mm | Hazardous to Aquatic Environment | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | 1 roll |
| 15654217 | 50 mm | Hazardous to Aquatic Environment | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | Pack of 250 |
| 15398751 | 50 mm | Hazardous to Aquatic Environment | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15664237 | 50 mm | Hazardous to Aquatic Environment | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15434861 | 70 mm | Hazardous to Aquatic Environment | Card Mounted | 6/Pk. | Pack of 6 |
| 15484851 | 100 mm | Health Hazard | Card Mounted | 20/Pk. | Pack of 4 |
| 11598884 | 100 mm | Health Hazard | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15624217 | 100 mm | Health Hazard | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15494851 | 40 mm | Health Hazard | Card Mounted | 20/Pk. | Pack of 20 |
| 15378751 | 50 mm | Health Hazard | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15654237 | 50 mm | Health Hazard | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 11783879 | 50 mm | Health Hazard | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | 1 roll |
| 15634217 | 50 mm | Health Hazard | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | Pack of 250 |
| 15404861 | 70 mm | Health Hazard | Card Mounted | 6/Pk. | Pack of 6 |
| 15424851 | 100 mm | Oxidizing | Card Mounted | 20/Pk. | Pack of 4 |
| 11598874 | 100 mm | Oxidizing | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15684207 | 100 mm | Oxidizing | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 11753879 | 50 mm | Oxidizing | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | 1 roll |
| 15694207 | 50 mm | Oxidizing | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | Pack of 250 |
| 15434851 | 40 mm | Oxidizing | Card Mounted | 20/Pk. | Pack of 20 |
| 15444861 | 100 mm | Respiratory Hazard | Card Mounted | 20/Pk. | Pack of 4 |
| 15358751 | 50 mm | Oxidizing | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15634237 | 50 mm | Oxidizing | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15444851 | 70 mm | Oxidizing | Card Mounted | 6/Pk. | Pack of 6 |
| 11598894 | 100 mm | Respiratory Hazard | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15664217 | 100 mm | Respiratory Hazard | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15454861 | 40 mm | Respiratory Hazard | Card Mounted | 20/Pk. | Pack of 20 |
| 11733889 | 50 mm | Respiratory Hazard | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | 1 roll |
| 15674217 | 50 mm | Respiratory Hazard | Continuous Roll, 250 Symbols (not labels)/Roll | 250/Pk. | Pack of 250 |
| 15308761 | 50 mm | Respiratory Hazard | Individual Markers on a Roll | 250/Pk. | 1 roll |
| 15674237 | 50 mm | Respiratory Hazard | Individual Markers on a Roll | 250/Pk. | Pack of 250 |
| 15464861 | 70 mm | Respiratory Hazard | Card Mounted | 6/Pk. | Pack of 6 |
| 11548874 | 100 mm | Flammable | Individual Markers on a Roll | 1 Ea. | 1 roll |
| 15664207 | 100 mm | Flammable | Individual Markers on a Roll | 1 Ea. | Pack of 250 |
| 11743879 | 50 mm | Flammable | Continuous Roll, 250 Symbols (not labels)/Roll | 1 Ea. | 1 roll |
| 15674207 | 50 mm | Flammable | Continuous Roll, 250 Symbols (not labels)/Roll | 1 Ea. | Pack of 250 |
| 15464841 | 40 mm | Explosive | Card Mounted | 20/Pk. | Pack of 20 |
| 15414851 | 70 mm | Flammable | Card Mounted | 6/Pk. | Pack of 6 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 72 options available.
This item is not returnable.
View return policy
Product Code.
15404851
Supplier
Brady™
Supplier No.
811685
Add to basket
11.12 EUR valid until 2026-10-31
BEST PRICE on Promo! Use promo code "28026" to get your promotional price.
BEST PRICE on Promo! Use promo code "28026" to get your promotional price.
Add to basket
This item is not returnable.
View return policy
Description
- Self-adhesive labels with GHS Symbol
- Laminated polyester with gloss finish
- High quality labels that can withstand the toughest environments
- Ultra-durable materials mean you only need to label products once in their lifetime
- Extremely weather, abrasion and chemical resistant
- Labels can withstand temperatures of -40 °C to +120 °C
- 50mm x 50mm labels are supplied as a continuous roll, user cuts as required
- 100mm x 100mm labels are individual labels on a roll
Specifica
Specifica
| Certifications/Compliance | ISO 7010, RoHS compliant using EU Directive 2002/95/EC |
| Alternate Part Number | PIC 1802-40*40-B7541-CRD |
| Finish | Gloss |
| Material | Polyester |
| Quantity | 20/Pk. |
| Style | Card Mounted |
| Adhesive Type | Permanent Acrylic |
| Description | Laminated Polyester |
| For Use With (Application) | Pipe markers and safety signs |
| Height (Metric) | 40 mm |
| Thickness | 0.024 mm |
| Width (Metric) | 40 mm |
| Temperature Range (Metric) | -40 °C-110 °C |
| Shape | Diamond |
| Displayed Text | Flammable |
| Product Type | GHS Symbol Sign |
| Color | Red on White |
| Vedi altri risultati Mostra meno risultati |
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?