missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester GHS Symbol Signs

Product Code. 15494851 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
40 mm
50 mm
70 mm
100 mm
Displayed Text:
Acute Toxicity
Compressed Gas
Corrosive
Explosive
Flammable
Hazardous to Aquatic Environment
Health Hazard
Oxidizing
Respiratory Hazard
Style:
Card Mounted
Continuous Roll, 250 Symbols (not labels)/Roll
Individual Markers on a Roll
Quantity:
1 Ea.
100/Pk.
20/Pk.
250/Pk.
6/Pk.
Unit Size:
1 roll
Pack of 20
Pack of 250
Pack of 4
Pack of 6
72 product options available for selection
Product selection table with 72 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Displayed Text Style Quantity Unit Size
15494851 40 mm Health Hazard Card Mounted 20/Pk. Pack of 20
15414871 40 mm Acute Toxicity Card Mounted 20/Pk. Pack of 20
15404871 100 mm Acute Toxicity Card Mounted 20/Pk. Pack of 4
11598904 100 mm Acute Toxicity Individual Markers on a Roll 250/Pk. 1 roll
15604227 100 mm Acute Toxicity Individual Markers on a Roll 250/Pk. Pack of 250
11753889 50 mm Acute Toxicity Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. 1 roll
15614227 50 mm Acute Toxicity Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. Pack of 250
15434871 70 mm Acute Toxicity Card Mounted 6/Pk. Pack of 6
15454851 100 mm Compressed Gas Card Mounted 20/Pk. Pack of 4
11548884 100 mm Compressed Gas Individual Markers on a Roll 250/Pk. 1 roll
15604217 100 mm Compressed Gas Individual Markers on a Roll 250/Pk. Pack of 250
15464851 40 mm Compressed Gas Card Mounted 100/Pk. Pack of 20
15368751 50 mm Compressed Gas Individual Markers on a Roll 250/Pk. 1 roll
15644237 50 mm Compressed Gas Individual Markers on a Roll 250/Pk. Pack of 250
15474851 70 mm Compressed Gas Card Mounted 6/Pk. Pack of 6
11763879 50 mm Compressed Gas Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. 1 roll
15614217 50 mm Compressed Gas Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. Pack of 250
15474861 100 mm Corrosive Card Mounted 20/Pk. Pack of 4
11548904 100 mm Corrosive Individual Markers on a Roll 250/Pk. 1 roll
15684217 100 mm Corrosive Individual Markers on a Roll 250/Pk. Pack of 250
15484861 40 mm Corrosive Card Mounted 20/Pk. Pack of 20
11743889 50 mm Corrosive Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. 1 roll
15694217 50 mm Corrosive Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. Pack of 250
15318761 50 mm Corrosive Individual Markers on a Roll 250/Pk. 1 roll
15684237 50 mm Corrosive Individual Markers on a Roll 250/Pk. Pack of 250
15328751 50 mm Explosive Individual Markers on a Roll 250/Pk. 1 roll
15614237 50 mm Explosive Individual Markers on a Roll 250/Pk. Pack of 250
15494861 70 mm Corrosive Card Mounted 6/Pk. Pack of 6
15338751 50 mm Flammable Individual Markers on a Roll 250/Pk. 1 roll
15624237 50 mm Flammable Individual Markers on a Roll 250/Pk. Pack of 250
15414861 100 mm Hazardous to Aquatic Environment Card Mounted 20/Pk. Pack of 4
11548894 100 mm Hazardous to Aquatic Environment Individual Markers on a Roll 250/Pk. 1 roll
15644217 100 mm Hazardous to Aquatic Environment Individual Markers on a Roll 250/Pk. Pack of 250
15424861 40 mm Hazardous to Aquatic Environment Card Mounted 20/Pk. Pack of 20
11723889 50 mm Hazardous to Aquatic Environment Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. 1 roll
15654217 50 mm Hazardous to Aquatic Environment Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. Pack of 250
15398751 50 mm Hazardous to Aquatic Environment Individual Markers on a Roll 250/Pk. 1 roll
15664237 50 mm Hazardous to Aquatic Environment Individual Markers on a Roll 250/Pk. Pack of 250
15434861 70 mm Hazardous to Aquatic Environment Card Mounted 6/Pk. Pack of 6
15484851 100 mm Health Hazard Card Mounted 20/Pk. Pack of 4
11598884 100 mm Health Hazard Individual Markers on a Roll 250/Pk. 1 roll
15624217 100 mm Health Hazard Individual Markers on a Roll 250/Pk. Pack of 250
15378751 50 mm Health Hazard Individual Markers on a Roll 250/Pk. 1 roll
15654237 50 mm Health Hazard Individual Markers on a Roll 250/Pk. Pack of 250
11783879 50 mm Health Hazard Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. 1 roll
15634217 50 mm Health Hazard Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. Pack of 250
15404861 70 mm Health Hazard Card Mounted 6/Pk. Pack of 6
15424851 100 mm Oxidizing Card Mounted 20/Pk. Pack of 4
11598874 100 mm Oxidizing Individual Markers on a Roll 250/Pk. 1 roll
15684207 100 mm Oxidizing Individual Markers on a Roll 250/Pk. Pack of 250
11753879 50 mm Oxidizing Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. 1 roll
15694207 50 mm Oxidizing Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. Pack of 250
15434851 40 mm Oxidizing Card Mounted 20/Pk. Pack of 20
15444861 100 mm Respiratory Hazard Card Mounted 20/Pk. Pack of 4
15358751 50 mm Oxidizing Individual Markers on a Roll 250/Pk. 1 roll
15634237 50 mm Oxidizing Individual Markers on a Roll 250/Pk. Pack of 250
15444851 70 mm Oxidizing Card Mounted 6/Pk. Pack of 6
11598894 100 mm Respiratory Hazard Individual Markers on a Roll 250/Pk. 1 roll
15664217 100 mm Respiratory Hazard Individual Markers on a Roll 250/Pk. Pack of 250
15454861 40 mm Respiratory Hazard Card Mounted 20/Pk. Pack of 20
11733889 50 mm Respiratory Hazard Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. 1 roll
15674217 50 mm Respiratory Hazard Continuous Roll, 250 Symbols (not labels)/Roll 250/Pk. Pack of 250
15308761 50 mm Respiratory Hazard Individual Markers on a Roll 250/Pk. 1 roll
15674237 50 mm Respiratory Hazard Individual Markers on a Roll 250/Pk. Pack of 250
15464861 70 mm Respiratory Hazard Card Mounted 6/Pk. Pack of 6
11548874 100 mm Flammable Individual Markers on a Roll 1 Ea. 1 roll
15664207 100 mm Flammable Individual Markers on a Roll 1 Ea. Pack of 250
11743879 50 mm Flammable Continuous Roll, 250 Symbols (not labels)/Roll 1 Ea. 1 roll
15674207 50 mm Flammable Continuous Roll, 250 Symbols (not labels)/Roll 1 Ea. Pack of 250
15404851 40 mm Flammable Card Mounted 20/Pk. Pack of 20
15464841 40 mm Explosive Card Mounted 20/Pk. Pack of 20
15414851 70 mm Flammable Card Mounted 6/Pk. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 72 options available.
72 options
This item is not returnable. View return policy
Product Code. 15494851 Supplier Brady™ Supplier No. 811700
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Help ensure safety at work and maximize productivity with these GHS warning labels. Brady™ Laminated Polyester GHS Symbol Signs are available in a variety of sizes, with and without text. Standardized images reduce the risk of misunderstandings.

  • Self-adhesive diamond shape labels
  • Pre-printed with GHS hazard symbols
  • Laminated polyester, gloss finish black and red on white

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number PIC 1805-40*40-B7541-CRD
Finish Gloss
Material Polyester
Quantity 20/Pk.
Style Card Mounted
Adhesive Type Permanent Acrylic, Pressure-sensitive
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 40 mm
Thickness 0.024 mm
Width (Metric) 40 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Displayed Text Health Hazard
Product Type GHS Symbol Sign
Color Black on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.