missing translation for 'onlineSavingsMsg'
Learn More

Lamivudine, 98%

Product Code. 15557049 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15557049 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557049 Supplier Thermo Scientific Acros Supplier No. 461090010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 134678-17-4 | C8H11N3O3S | 229.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 134678-17-4
Melting Point 160.0°C to 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C8H11N3O3S
Quantity 1 g
Synonym lamivudine, epivir, zeffix, heptovir, epivir-hbv, hepitec, heptodin, 2',3'-dideoxy-3'-thiacytidine, lamivir, 3tc
Solubility Information Solubility in water: 70g/L (20°C).
InChI Key JTEGQNOMFQHVDC-NKWVEPMBSA-N
SMILES C1C(OC(S1)CO)N2C=CC(=NC2=O)N
IUPAC Name 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
Molecular Weight (g/mol) 229.26
PubChem CID 60825
ChEBI CHEBI:63577
Formula Weight 229.26
Percent Purity 98%
Chemical Name or Material Lamivudine
Show More Show Less
compliance-icons
Product Identifier
  • Lamivudine
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.