missing translation for 'onlineSavingsMsg'
Learn More

Lamivudine, TRC

Product Code. 18000310
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18000310 100 mg 100mg
18071000 50 mg 50mg
17959819 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18000310 Supplier Toronto Research Chemicals Supplier No. TRCL172500100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134678-17-4
Molecular Formula C8 H11 N3 O3 S
Molecular Weight (g/mol) 229.26
Synonym Lamivudine, 2(1H)-Pyrimidinone, 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, 2(1H)-Pyrimidinone, 4-amino-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, (2R-cis)-, 1,3-Oxathiolane, 2(1H)-pyrimidinone deriv., (-)-2'-Deoxy-3'-thiacytidine, (-)-BCH 189, 2-Hydroxymethyl-5-(cytosin-1-yl)-1,3-oxathiolane, 3TC, BCH 189, (-)-, Epivir, Epivir HBV, GR 109714X, Hepitec, Heptodin, Heptovir, L-SddC, Lamivir, Virolam, Zeffix, Zefix, cis(-)-2-Hydroxymethyl-5-(cytosin-1-yl)-1,3-oxathiolane, β-L-2',3'-Dideoxy-3'-thiacytidine, β-L-3'-Thia-2',3'-dideoxycytidine
IUPAC Name 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@@H]2CS[C@H](CO)O2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H11N3O3S/c9-5-1-2-11(8(13)10-5)6-4-15-7(3-12)14-6/h1-2,6-7,12H,3-4H2,(H2,9,10,13)/t6-,7+/m0/s1
Formula Weight 229.0521
Chemical Name or Material Lamivudine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.