Learn More
UVP Lamp XX-40S bench brackets included aluminum 220-240V 50/60Hz length 1260mm supplied with 2 x 254nm 40W tubes UVP
Product Code. 11709064
Change view
Click to view available options
Voltage:
220/240 V
Unit Size:
Each
Product Code. Voltage Unit Size
11709064 220/240 V Each
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11709064 Supplier UVP Supplier No. 95004308
  • ONLINE EXCLUSIVE PRICE
    929.86 €
    878.45 € / Pack of 1
    Save 51.41 €
    5% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below