missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole, MedChemExpress

Product Code. 16471369
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471369 1 g 1g
16468467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471369 Supplier Medchem Express Supplier No. HY13662
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lansoprazole (AG 1749) is an orally active proton pump inhibitor which prevents the stomach from producing acid. Lansoprazole (AG 1749) is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).

Lansoprazole (AG 1749) is an orally active proton pump inhibitor which prevents the stomach from producing acid. Lansoprazole (AG 1749) is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).

Specifications

Chemical Name or Material Lansoprazole
CAS 103577-45-3
Color White
Purity Grade Notes Research
Molecular Formula C16H14F3N3O2S
Quantity 1 g
Synonym AG-1749
Solubility Information DMSO : 100 mg/mL (270.74 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.27 mM; Need ultrasonic)
SMILES O=S(C1=NC2=CC=CC=C2N1)CC3=NC=CC(OCC(F)(F)F)=C3C
Molecular Weight (g/mol) 369.36
Formula Weight 369.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.