missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole N-Oxide, TRC

Product Code. 30134478
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134478 25 mg 25mg
30008379 10 mg 10mg
30007126 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134478 Supplier Toronto Research Chemicals Supplier No. TRCL17503525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213476-12-1
Molecular Formula C16 H14 F3 N3 O3 S
Molecular Weight (g/mol) 385.36
Synonym 2-[(RS)-[[3-Methyl-1-oxido-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methyl]sulphinyl]-1H-benzimidazole, Lansoprazole Imp. A (EP), Lansoprazole N-Oxide
IUPAC Name 2-[[3-methyl-1-oxido-4-(2,2,2-trifluoroethoxy)pyridin-1-ium-2-yl]methylsulfinyl]-1H-benzimidazole
SMILES Cc1c(OCC(F)(F)F)cc[n+]([O-])c1CS(=O)c2nc3ccccc3[nH]2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H14F3N3O3S/c1-10-13(22(23)7-6-14(10)25-9-16(17,18)19)8-26(24)15-20-11-4-2-3-5-12(11)21-15/h2-7H,8-9H2,1H3,(H,20,21)
Formula Weight 385.0708
Percent Purity >95%
Chemical Name or Material 2-[(RS)-[[3-Methyl-1-oxido-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methyl]sulphinyl]-1H-benzimidazole (Lansoprazole N-Oxide)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.