missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole Sulfone, TRC

Product Code. 30008881
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008881 10 mg 10mg
30004487 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008881 Supplier Toronto Research Chemicals Supplier No. TRCL17502610MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131926-99-3
Molecular Formula C16 H14 F3 N3 O3 S
Molecular Weight (g/mol) 385.36
Synonym 2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methyl]sulphonyl]-1H-benzimidazole, Lansoprazole Imp. B (EP), Lansoprazole Sulphone, 1H-Benzimidazole, 2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfonyl]- (9CI), 2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfonyl]-1H-benzimidazole, AG 1813, Lansoprazole sulfone
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfonyl]-1H-benzimidazole
SMILES Cc1c(CS(=O)(=O)c2nc3ccccc3[nH]2)nccc1OCC(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H14F3N3O3S/c1-10-13(20-7-6-14(10)25-9-16(17,18)19)8-26(23,24)15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)
Formula Weight 385.0708
Percent Purity >95%
Chemical Name or Material Lansoprazole Sulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.