missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 100 Orientation, Thermo Scientific™

Product Code. 11354148
Click to view available options
Quantity:
1 Ea.
5 Ea.
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 11354148

Brand: Thermo Scientific Alfa Aesar 041434.KU

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

100 Orientation

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12003-65-5
Molecular Formula LaAlO3
MDL Number MFCD00144550

Specifications

Color Brown-Yellow
Physical Form Plate
Quantity 5 Ea.
Odor Odorless
Chemical Name or Material Lanthanum Aluminum Oxide Substrate, Polished Two Sides
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.