missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride, 99.9%

Product Code. 10287660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
Product Code. Quantity Packaging unitSize
10287660 10 g Glass bottle 10g
10404545 50 g Glass bottle 50g
13448518 250 g Glass bottle 250g
3 options
This item is not returnable. View return policy

Product Code. 10287660

Brand: Thermo Scientific Acros 315230100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10099-58-8 | Cl3La | 245.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Boiling Point 1000.0°C
Melting Point 860.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9%
Linear Formula LaCl3
Merck Index 15,5414
Solubility Information Solubility in water: soluble
Formula Weight 245.26
Percent Purity 99.9%
Packaging Glass bottle
Chemical Name or Material Lanthanum chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lanthanum chloride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Lanthanum chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.