missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride heptahydrate, 99.99%, (trace metal basis)

Product Code. 10762621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762621 100 g Glass bottle 100g
10125403 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10762621 Supplier Thermo Scientific Acros Supplier No. 193271000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Lanthanum chloride heptahydrate, 99.99%, Cl3La.7H2O, CAS Number-10025-84-0, 500g, White, 371.38, Trace Metal Basis, GHS Signal Word: Danger | CAS: 10025-84-0 | Cl3H14LaO7 | 371.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]

Specifications

Melting Point 91.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 99.99%
Linear Formula LaCl3·7H2O
Merck Index 15, 5414
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acids and alcohols
Formula Weight 371.38
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lanthanum chloride heptahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lanthanum chloride heptahydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Lanthanum chloride heptahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.