missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride, Releasing Agent Solution, Specpure™

Product Code. 15456295
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456295 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456295 Supplier Thermo Scientific Alfa Aesar Supplier No. 039036.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10099-58-8 | Cl3La | 245.26 g/mol

Lanthanum chloride is a precursor for synthesis of lanthanum phosphate nano rods and used in gamma detectors. It is also used as a catalyst for high pressure oxidative chlorination of methane to chloromethane with hydrochloric acid and oxygen. In organic synthesis, lanthanum trichloride acts as lewis acid for the conversion of aldehydes to acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10099-58-8
Physical Form Liquid
Quantity 500 mL
Molecular Formula Cl3La
MDL Number MFCD00011068
Solubility Information Miscible with water.
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[La+3]
IUPAC Name lanthanum(3+) trichloride
Molecular Weight (g/mol) 245.26
Chemical Name or Material Lanthanum chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.