missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) 2-methoxyethoxide, 99.9% (REO), 5% w/v in 2-methoxyethanol

Product Code. 11319098
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 11319098

Brand: Thermo Scientific Alfa Aesar 042522.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Preparation of lanthanum oxide thin films by the Sol-Gel method | CAS: 115685-56-8

Lanthanum(III) 2-methoxyethoxide is used in the preparation of lanthanum oxide thin films by the Sol-Gel method

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115685-56-8
MDL Number MFCD00210619
Synonym lanthanum iii 2-methoxyethoxide, lanthanum tris 1-methoxyethan-1-olate, acmc-20akoi, lanthanum 3+ ion tris 1-methoxyethanolate

Specifications

Color Yellow
Density 0.96 g/mL
Boiling Point 124°C
Flash Point 40°C (104°F)
Linear Formula La(OCH2CH2OCH3)3
Quantity 100 mL
UN Number UN1188
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 364.17
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Lanthanum (III) 2-methoxyethoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.