missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) carbonate hydrate, REacton™, 99.99% (REO)

Product Code. 11312298
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312298 250 g 250g
11302298 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312298 Supplier Thermo Scientific Alfa Aesar Supplier No. 013197.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54451-24-0 | C3La2O9 | 457.84 g/mol

Lanthanum(III) carbonate hydrate is used as a starting material for the preparation of lanthanum strontium manganite, which is used in solid oxide fuel cell applications. It is also involved in the preparation of high-temperature superconductors. It finds application in medicine as a phosphate binder. Further, it is used in the treatment of hyperphosphatemia. In addition to this, it is used for the tinting of glass, water treatment and as a catalyst for hydrocarbon cracking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54451-24-0
Molecular Formula C3La2O9
Molecular Weight (g/mol) 457.84
MDL Number MFCD00011069
InChI Key NZPIUJUFIFZSPW-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) tricarbonate
SMILES [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Lump/Powder
Quantity 250 g
Merck Index 14,5364
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 457.85 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Density 2.6 g/mL to 2.7 g/mL
Assay (REO)
Chemical Name or Material Lanthanum(III) carbonate hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Lanthanum(III) carbonate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Lanthanum(III) carbonate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.