missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%, Solstice

Product Code. 15603520
Change view
Click to view available options
Quantity:
1 Ea.
100 g
6 x 1 Ea.
6 x 100 g
Packaging:
Ampoule
Poly bottle
Unit Size:
100g
Each
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15603520 100 g Poly bottle 100g
15613520 6 x 100 g Poly bottle Pack of 6
15633650 1 Ea. Ampoule Each
15643650 6 x 1 Ea. Ampoule Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15603520 Supplier Honeywell-Fluka Supplier No. 31820100G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
This item is not returnable. View return policy

for 1L standard solution

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0

Specifications

Quantity 100 g
Molecular Formula Cl3H14LaO7
Linear Formula LaCl3 · 7H2O
MDL Number MFCD00149756
UN Number NONH for all modes of transport
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
IUPAC Name lanthanum(3+) heptahydrate trichloride
Molecular Weight (g/mol) 371.36
Formula Weight 371.37g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Packaging Poly bottle
Chemical Name or Material Lanthanum(Iii) Chloride Heptahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lanthanum(III) chloride heptahydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Lanthanum chloride heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.