missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%, Solstice

Product Code. 15613520
Change view
Click to view available options
Quantity:
1 Ea.
100 g
6 x 1 Ea.
6 x 100 g
Packaging:
Ampoule
Poly Bottle
Unit Size:
100g
Each
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15613520 6 x 100 g Poly Bottle Pack of 6
15603520 100 g Poly Bottle 100g
15633650 1 Ea. Ampoule Each
15643650 6 x 1 Ea. Ampoule Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15613520 Supplier Honeywell-Fluka Supplier No. 318206X100G
  • ONLINE EXCLUSIVE PRICE
    665.00 €
    600.42 € / Pack of 6
    Save 64.58 €
    9% Off
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 1L standard solution

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0

Specifications

Melting Point 91°C (decomposition)(literature)
CAS Max % ≤100.0000%
Physical Form Solid
Quantity 6 x 100 g
Molecular Formula Cl3H14LaO7
Linear Formula LaCl3 · 7H2O
MDL Number MFCD00149756
UN Number UN3260
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
IUPAC Name lanthanum(3+) heptahydrate trichloride
Molecular Weight (g/mol) 371.36
Formula Weight 371.37
Percent Purity >99%
Grade puriss. p.a.
Packaging Poly Bottle
Chemical Name or Material Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.