missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%, Solstice

Product Code. 15633650
Change view
Click to view available options
Quantity:
1 Ea.
100 g
6 x 1 Ea.
6 x 100 g
Packaging:
Ampoule
Poly bottle
Unit Size:
100g
Each
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15633650 1 Ea. Ampoule Each
15603520 100 g Poly bottle 100g
15613520 6 x 100 g Poly bottle Pack of 6
15643650 6 x 1 Ea. Ampoule Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15633650 Supplier Honeywell-Fluka Supplier No. 320381EA
  • ONLINE EXCLUSIVE PRICE
    67.12 €
    63.16 € / Pack of 1
    Save 3.96 €
    5% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 1L standard solution

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710

Specifications

Physical Form Liquid
Quantity 1 Ea.
Packaging Ampoule
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
UN Number NONH for all modes of transport
Beilstein 3534976
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Formula Weight 58.44g/mol
Grade Volumetric
Chemical Name or Material Sodium Chloride Concentrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.