missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) nitrate hexahydrate, Solstice

Product Code. 15646040
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15646040 500 g 500g
15636040 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15646040 Supplier Honeywell-Fluka Supplier No. 61520500G
  • ONLINE EXCLUSIVE PRICE
    930.00 €
    836.45 € / 500g
    Save 93.55 €
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a.,≥99.0% (titration)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate
PubChem CID 16211478
IUPAC Name lanthanum(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 126°C
Melting Point 40°C
Quantity 500 g
Appearance Crystalline
Assay Percent Range ≥99.0%
Linear Formula La(NO3)3 · 6H2O
UN Number UN1477
Solubility Information 1580 g/L in Water
Formula Weight 433.01g/mol
Percent Purity ≥99% (titration)
Grade ACS
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.