missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, 91%

Product Code. 10035710 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
This item is not returnable. View return policy

Product Code. 10035710

Brand: Thermo Scientific Acros 413021000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum(III) oxide, 91%, La2O3, CAS Number-1312-81-8, 100g, 89% min, 4200.0 deg.C, White to Gray, 215-200-5, 325.82, HYGROSCOPIC, GHS H Statement: Causes serious eye irritation. | CAS: 1312-81-8 | La2O3 | 325.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00011071

Specifications

Boiling Point 4200.0°C
Melting Point 2315.0°C
Color White to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 89% min
Linear Formula La2O3
Merck Index 15, 5414
Solubility Information Solubility in water: insoluble.
Formula Weight 325.82
Percent Purity 91%
Packaging Glass bottle
Chemical Name or Material Lanthanum(III) oxide, 91%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado