missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Amine Reactive Tb Chelate

Product Code. 11563467
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 11563467

Brand: Invitrogen™ PV3583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development.

As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development. The amine-reactive, isothiocyanate group conjugates to virtually any peptide or protein that contains one or more accessible amine moieties, such as the amine group found on a lysine residue or an unmodified N-terminal amine group of a peptide or protein. Amine-modified DNA oligonucleotides can be labeled as well. Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity. See the user guide for application-based protocols for labeling peptides containing free amino groups, amine-modified oligonucleotides, and IgG antibodies.

TRUSTED_SUSTAINABILITY

Specifications

Description LanthaScreen™ Amine Reactive Tb Chelate, 10μg, 1050.88g/mol Molecular Weight, ≥90% by HPLC Purity, Store at –20°C, Used For Pharma and Biopharma, Target and Lead Identification and Validation
For Use With (Application) Drug discovery & development, target & lead identification & validation, TR-FRET assay development
Quantity 10 μg
Product Type Amine Reactive Tb Chelate
Kit Contents LanthaScreen™ TR-FRET labeling reagents are provided lyophilized
Molecular Weight (g/mol) 1050.88g/mol
Purity ≥90% by HPLC
Content And Storage LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day.
Shipping Condition Dry Ice
Product Line LanthaScreen

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.