Learn More
Invitrogen™ LanthaScreen™ Eu-Streptavidin
LanthaScreen™ Eu-Streptavidin
Brand: Invitrogen™ PV6025
Description
A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates.Specifications
See the Certificate of Analysis | |
55kDa | |
10mM HEPES (pH 7.5), 137mM NaCl and 2.7mM KCl (HEPES buffered saline) | |
Frozen | |
10mM HEPES (pH 7.5), 137mM NaCl and 2.7mM KCl (HEPES buffered saline) |
LanthaScreen™ Eu-Streptavidin, 1mg, 0.25mg/mL Concentration, Frozen, 55kDa Molecular Weight, Alexa Fluor™ 647, Europium Dye, Store at –20°C, 340/615, 665nm Excitation/Emission Wavelength, Used For Target & Lead Identification & Validation | |
1 mg | |
-20°C | |
Pharma and biopharma, target and lead identification and validation | |
LanthaScreen™ Eu-Streptavidin |
For Research Use Only. Not for use in diagnostic procedures.