missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Eu-Streptavidin

Product Code. p-7076006
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10630910

Brand: Invitrogen™ PV5899

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LanthaScreen™ Eu-Streptavidin

A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates.

TRUSTED_SUSTAINABILITY

Specifications

Concentration See the Certificate of Analysis
Conjugate Alexa Fluor 647, Europium
Quantity 25 μg
Content And Storage Europium-labeled Streptavidin in HEPES buffered saline. Store at -20°C.
Assay Entry Biochemical binding/kinetic
Excitation/Emission 340/615, 665 nm
Shipping Condition Dry Ice
Target Entry ABL1 G250E, STK16, ZAP70, MAP2K1 S218D S222D, MAPK8, CAMK2B, CHEK1, STK33, IGFR1, BLK, LYNA, NEK2, CHUK, MAPKAPK5, MAP3K7/MAP3K7IP1, DAPK2, HIPK3, MAP2K1, MAP2K6, PIK3CG, ABL1 E255K, BMX, MAP4K2, MAPK9, CAMK1D, CHEK2, PIM1, STK22D, CSNK1G2, EPHA3, EPHB4, LCK, YES1, MET, PRKCI, WNK3, ABL1, ABL2, FGFR3, FGFR3 K650E, KIT V654A, AURKB, CDC42BPA, CDK1, CDK8, CDK9/Cyclin K, LYNB, CDC42BPB, CDK2, CDK7, PDGFRB, MET M1250T, RPS6KA1, LIMK1, MAP2K2, PIK3CA/PIK3R1, MAP2K6 S207E T211E, SRC, ABL1 Y253F, KIT T670I, RPS6KA2, PAK7, IRAK4, CSK
Form Liquid
Product Line LanthaScreen
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye Alexa Fluor 647, Europium
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.