missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ LRRK2 [pSer935] Cellular Assay Kit

Product Code. 13453269
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 13453269

Brand: Thermo Scientific™ A14685

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The LanthaScreen™ LRRK2 [pSer935] Cellular Assay Kit combines the flexibility of the BacMam gene delivery system with the robustness and power of our LanthaScreen™ TR-FRET technology.

The LanthaScreen™ LRRK2 [pSer935] Cellular Assay Kit combines the flexibility of the BacMam gene delivery system with the robustness and power of our LanthaScreen™ TR-FRET technology. It allows researchers to interrogate phosphorylation at serine 935 on a wild type Leucine-Rich Repeat Kinase-2 (LRRK2) protein in a variety of cell backgrounds to identify LRRK2 kinase inhibitors.

This assay format provides a convenient tool for the expression of LRRK2-GFP fusion protein in the cell background of of your choice. The resulting cellular assay can be used to identify LRRK2 and mutant inhibitorss, aiding in Parkinson's disease research and other research areas.

This kit provides enough material to evaluate the technology in a 384-well plate; larger sizes of all reagents are available separately.

The kit includes:

  • LanthaScreen™ Tb-anti-LRRK2 [pSer935] Antibody, 5 μg
  • BacMam LRRK2-GFP Wild Type Reagent, 5 x 1 mL
  • LanthaScreen™ 6X Lysis Buffer, 6 mL
  • Instrument Control Terbium TR-FRET Kit

A kit for interrogating a mutant LRRK2 protein (Cat. No. A14686) is also available.

Benefits of this assay include:

  • More Physiologically Relevant—cell-based LRRK2 kinase activity and choice of cell background
  • More Convenient—compatible with High Throughput Screening (HTS); LanthaScreen™ technology reduces interference and noise
  • Disease Relevant—screen compound libraries for Parkinson's disease-relevant molecules

More Physiologically Relevant

The LanthaScreen™ Tb-Anti-LRRK2 [pSer935] Antibody allows the investigation of phosphorylation at serine 935 on LRRK2 protein kinase. When paired with LRRK2 BacMam reagents, you have the freedom to choose the cellular background for your assay, including primary cells. This enables screening for potential inhibitors of LRRK2 proteins in their natural complexes in a physiologically relevant cell type.

More Convenient

Assays are run in a fully homogenous, addition-only format without any of the washing, lysate transfer, or separation procedures required for traditional methods such as western blotting and ELISA, making this assay ideal for HTS applications. The LanthaScreen™ technology provides all of the advantages of TR-FRET ratiometric detection, including reduced data noise, less interference from fluorescent compounds, and high sensitivity, which results in the use of fewer cells than traditional Western or ELISA methods.

Disease Relevant

The LanthaScreen™ Tb-Anti-LRRK2 [pSer935] Antibody can be used by Parkinson's and other disease researchers to perform HTS, cell-based assays to generate inhibition curves (IC50) for their compound libraries. This will aid drug discovery efforts by reducing the time to results from 4 hrs to typically 1.5 hr. compared to alternative ELISAs.

TRUSTED_SUSTAINABILITY

Specifications

Assay Cell-Based Assay
Assay Entry Cell-based terbium modification activity (transient)
Cell State Cells Not Included
Content And Storage • BacMam LRRK2-GFP, 5 x 1 mL (Store at 4°C)
• LanthaScreen™ 6X Lysis Buffer, 1 tube (Store at 4°C)
• LanthaScreen™ Tb-anti-LRRK2 [pSer935] Antibody, 1 tube (Store at -20°C)
• Instrument Control Tb TR-FRET Kit (Store at 4°C)
• LanthaScreen™ LRRK2 [pSer935] Cellular Assay Kit Protocol (Store at room temperature)
Description LanthaScreen™ LRRK2 [pSer935] Cellular Assay Kit, 1 Kit, Fluorescent Detection Method, Kinases Druggable Target, LanthaScreen™ System, TR-FRET (Activity) Readout, Room Temperature Storage
Detection Method Fluorescence
For Use With (Application) TR-FRET
For Use With (Equipment) Microplate Reader
Includes 5 tubes BacMam LRRK2-GFP 1mL, Store at 4°C, 1 tube LanthaScreen™ 6X Lysis Buffer, Store at 4°C, 1 Tube LanthaScreen™ Tb-anti-LRRK2 [pSer935] Antibody, Store at –20°C, 1 Instrument Control Tb TR-FRET Kit, Store at 4°C, 1 LanthaScreen™ LRRK2 [pSer935] Cellular Assay Kit Protocol, Store at room temperature
Product Line LanthaScreen
Quantity 1 kit
Readout TR-FRET (Activity)
Reporter Gene GFP (EmGFP)
Target Entry LRRK2
Type Cellular Assay Kit
System Type LanthaScreen™
Druggable Target Kinases
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.