missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-labeled Anti-Goat Ab

Product Code. 10666255
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10666255 1 mg Each
10369244 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10666255 Supplier Thermo Scientific™ Supplier No. PV3772
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is ideally suited for use during assay development when evaluating multiple primary goat antibodies, or when it is not possible to directly terbium label a primary antibody.

  • A general reagent for use in assay development, this terbium labeled antibody will bind goat antibodies
  • Ideally suited for use during assay development when evaluating multiple primary goat antibodies
  • Also for use when it is not possible to to directly terbium label a primary antibody
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Terbium-labeled anti-Goat antibody in HEPES buffered saline. Store at -20°C.
Excitation/Emission 340/490, 520
Readout End Point
Shipping Condition Dry Ice
Conjugate Lanthanide Chelates, Terbium (Tb)
Product Line LanthaScreen
Quantity 1 mg
Reactivity Goat

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.