missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-labeled Anti-Goat Antibody Kit

Product Code. 11503527
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11503527 25 μg 25µg
10779544 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11503527 Supplier Thermo Scientific™ Supplier No. PV3769
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents to facilitate in-house assay development around multiple target classes and assay formats.

  • Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents to facilitate in-house assay development around multiple target classes and assay formats
  • Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species
  • Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control
  • In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled
  • The detection step is performed by the addition of the Tb-labeled secondary antibody
  • This approach can be applied to other target classes and assay formats
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Dry Ice
Conjugate Lanthanide Chelates, Terbium (Tb)
Product Line LanthaScreen
Quantity 25 μg
Reactivity Goat

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.