missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-labeled Anti-rabbit Ab

Product Code. 10666445
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10666445 1 mg Each
10359194 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10666445 Supplier Invitrogen™ Supplier No. PV3776
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A general reagent for use in assay development, this terbium labeled antibody will bind rabbit antibodies.

A general reagent for use in assay development, this terbium labeled antibody will bind rabbit antibodies. This product is ideally suited for use during assay development when evaluating multiple primary rabbit antibodies or when you are unable to directly terbium label your primary antibody.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Terbium-labeled anti-Rabbit antibody in HEPES buffered saline.

Store at -20°C.
Excitation/Emission 340/490, 520 nm
Readout End Point
Shipping Condition Dry Ice
Conjugate Lanthanide Chelates, Terbium (Tb)
Product Line LanthaScreen
Quantity 1 mg
Reactivity Rabbit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.