missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Tb-Streptavidin, 1 mg/mL Kit
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
Description
As part of the LanthaScreen™ TR-FRET Toolbox of assay reagents, LanthaScreen™ Tb-labeled streptavidin is a generic reagent that can be used to easily label biotinylated biomolecules with terbium for use in TR-FRET assays. Although the terbium chelate donor can be directly attached to a biomolecule such as a protein or peptide, the Tb-labeled streptavidin can be indirectly attached to a target molecule through a biotin-mediated association with Tb-labeled streptavidin.
Specifications
Specifications
| Concentration | 1 mg/mL |
| Conjugate | Tb (Terbium) |
| Quantity | 50 μg |
| Content And Storage | LanthaScreen™ Tb-Streptavidin is supplied at 1 mg/ml and contains the streptavidin in HEPES buffered saline, 400 μl of the fluorescein-labeled, biotinylated peptide for use as a positive control, and 100 ml of TR-FRET Dilution Buffer. Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C). |
| Shipping Condition | Dry Ice |
| Form | Liquid |
| Product Line | LanthaScreen |
| Product Type | Streptavidin Conjugate (Fluorescent) |
| Label or Dye | Tb (Terbium) |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction