missing translation for 'onlineSavingsMsg'
Learn More

LanthaScreen™ Tb-Ubiquitin

Product Code. 11515016
Click to view available options
Quantity:
25 μg
5 μg
Unit Size:
25µg
5µg
This item is not returnable. View return policy

Product Code. 11515016

Brand: Invitrogen PV4376

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LanthaScreen™ Tb-Ubiquitin

Ubiquitin has been labeled with the LanthaScreen™ Terbium chelate for use in high throughput assays to monitor protein ubiquitination. Due to the method used to label the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.

For use in combination with FRET acceptors including Fluorescein-ubiquitin (PV4377) or a fluorescein-labeled antibody.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 μg
Conjugate Terbium (Tb)
Content And Storage Terbium-labeled ubiquitin in HEPES buffered saline. Store at -20°C.
Detection Method Fluorescence
Excitation/Emission 340⁄490, 520
For Use With (Equipment) Microplate Reader
Label or Dye Tb (Terbium)
Label Type Lanthanide Chelates
Readout End Point
Shipping Condition Dry Ice
Substrate Properties Protein-Based Substrate
Technique TR-FRET
Form Liquid
Product Line LanthaScreen™
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.