missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit

Product Code. 15617858
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15617858 800 x 20 μL assays Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15617858 Supplier Thermo Scientific™ Supplier No. A15885
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential estrogen receptor (ER) alpha ligands as agonists or antagonists of ligand-dependent coactivator recruitment.

The LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit uses an ER alpha ligand-binding domain (ER alpha-LBD) tagged with glutathione-S-transferase (GST), a terbium (Tb)-labeled anti-GST antibody, and a fluorescein-labeled coactivator peptide in a homogenous mix-and-read assay format.

Agonist Mode

To run the LanthaScreen TR-FRET ER alpha Coactivator Assay in agonist mode (to identify agonist compounds), ER alpha-LBD is added to ligand test compounds, followed by the addition of a mixture of fluorescein-coactivator peptide and Tb-anti-GST antibody. After incubation at room temperature, the TR-FRET 520:495 nm emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound. Based on the biology of the ER alpha-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide.

Antagonist Mode

To run the LanthaScreen TR-FRET ER alpha Coactivator Assay in antagonist mode (to identify antagonist compounds), ER alpha-LBD is added to ligand test compounds, followed by addition of a mixture of agonist, fluorescein-coactivator peptide, and terbium (Tb)-labeled anti-GST antibody. The concentration of agonist used in this mode is the EC80 concentration as determined by first running the assay in agonist mode.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 tube ER alpha Ligand Binding Domain Recombinant Protein (store at -68 to -85°C)
1 tube of Peptide (store at -5 to -30°C)
2 bottles TR-FRET Buffer (store at -5 to -30°C)
1 tube Tb-Anti-GST Ab (store at -5 to -30°C)
1 tube of DTT (store at -5 to -30°C)
For Use With (Application) Co-Factor Interaction Assay, Nuclear Receptor Assay
Product Line LanthaScreen
Quantity 800 x 20 μL assays
Type Coactivator Assay Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.