missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ TR-FRET ERR gamma Coactivator Assay Kit, goat

Product Code. 10585813
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10585813 800 x 20 μL assays Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10585813 Supplier Thermo Scientific™ Supplier No. PV4408
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The LanthaScreen™ TR-FRET Estrogen Related Receptor (ERR) gamma Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential ERR gamma ligands as antagonists/inverse agonists of coactivator displacement.

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15121:

The LanthaScreen™ TR-FRET Estrogen Related Receptor (ERR) gamma Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential ERR gamma ligands as antagonists/inverse agonists of coactivator displacement. The homogeneous mix-and-read assay format results in a ligand EC50 composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and displace coactivator peptide (Figure 1).

How it works

The LanthaScreen™ TR-FRET ERR gamma Coactivator Assay Kit includes a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human ERR gamma ligand-binding domain (ERR gamma-LBD) that is tagged with glutathione-S-transferase (GST). To assay, ERR gamma-LBD is added to test compounds, followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-anti- GST antibody. After room temperature incubation, the TR-FRET 520:495 emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound.

Contents and Storage:

The LanthaScreen™ TR-FRET ERR gamma Coactivator Assay Kit contains ERR gamma-LBD (GST) protein, fluorescently labeled PGC1a coactivator peptide, Tb-anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)
Description LanthaScreen™ TR-FRET ERR Gamma Coactivator Assay Kit, 800 x 20μL Assays, 384-Well Plate Format, Tb (Terbium) Dye Type, Fluorescent Detection Method, TR-FRET Technique, For Pharma & Biopharma, Target & Lead Identification & Validation
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Includes ERR gamma-LBD (GST) protein, fluorescently labeled PGC1a coactivator peptide, Tb-anti-GST antibody, and buffers
Quantity 800 x 20 μL assays
Regulatory Status For research use only (RUO)
Packaging 384-well Plate
Content And Storage Store in ultra-cold freezer (-68°C to -85°C).
Assay Entry Biochemical coregulator interaction
Gene ID (Entrez) 2104
Readout End Point
Shipping Condition Dry Ice
Target Entry ERR3, ESRRG, NR3B3
Product Line LanthaScreen
For Use With (Application) Co-Factor Interaction Assay, TR-FRET
Ligand ERR gamma
Product Type TR-FRET ERR Gamma Coactivator Assay Kit
No. of Assays 800 x 20 μL Assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.