missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ TR-FRET LXR alpha Coactivator Assay Kit, goat

Product Code. 10413273
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Pack of 800
This item is not returnable. View return policy

Product Code. 10413273

Brand: Invitrogen™ PV4655

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LanthaScreen™ TR-FRET LXR alpha Coactivator Assay Kit, goat

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15128:

The LanthaScreen™ TR-FRET LXR (liver X receptor) alpha Coactivator Assay provides a sensitive and robust method for high-throughput screening of potential LXR alpha ligands as agonists of coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and an LXR alpha ligand-binding domain (LBD) that is tagged with glutathione-S-transferase (GST), in a homogeneous mix-and-read assay format.

In the LanthaScreen™ TR-FRET LXR alpha Coactivator Assay, LXR alpha-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the LXR alpha-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).

Contents and Storage:
The LanthaScreen™ TR-FRET LXR alpha Coactivator Assay Kit contains LXR alpha-LBD (GST) protein, fluorescently labeled TRAP220/DRIP-2 coactivator peptide, Tb-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Quantity 800 x 20 μL assays
Packaging 384-well Plate
Content And Storage Store in ultra-cold freezer (-68 to -85°C).
Assay Entry Biochemical coregulator interaction
Gene ID (Entrez) 10062
Readout End Point
Shipping Condition Dry Ice
Target Entry LXRA, LXR alpha, NR1H3
Product Line LanthaScreen
For Use With (Application) Co-Factor Interaction Assay, TR-FRET
Ligand LXR alpha
Product Type TR-FRET LXR Alpha Coactivator Assay Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.