missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ TR-FRET LXR alpha Coactivator Assay Kit, rabbit

Product Code. 15313796
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15313796 800 x 20 μL assays 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15313796 Supplier Thermo Scientific™ Supplier No. A15128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The LanthaScreen™ TR-FRET LXR (liver X receptor) alpha Coactivator Assay provides a sensitive and robust method for high-throughput screening of potential LXR alpha ligands as agonists of coactivator recruitment.

This kit contains Rabbit Tb-Anti-GST antibody; other kit components are the same as kit PV4655:

The LanthaScreen™ TR-FRET LXR (liver X receptor) alpha Coactivator Assay provides a sensitive and robust method for high-throughput screening of potential LXR alpha ligands as agonists of coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and an LXR alpha ligand-binding domain (LBD) that is tagged with glutathione-S-transferase (GST), in a homogeneous mix-and-read assay format.

In the LanthaScreen™ TR-FRET LXR alpha Coactivator Assay, LXR alpha-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the LXR alpha-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).

TRUSTED_SUSTAINABILITY

Especificaciones

Color Green
Conjugate Tb (Terbium)
Description LanthaScreen™ TR-FRET LXR Alpha Coactivator Assay Kit, 800 x 20μL Assays, Green, 384-Well Plate Format, Tb (Terbium) Dye Type, Fluorescent Detection Method, TR-FRET Technique, For Pharma & Biopharma, Target & Lead Identification, Validation
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Includes Tb-anti-GST antibody (rabbit), LXR alpha-LBD, GST, fluorescein coactivator peptide, TR-FRET coregulator buffer H, 1M DTT
Quantity 800 x 20 μL assays
Regulatory Status For research use only (RUO)
Packaging 384-well Plate
Content And Storage • Tb-anti-GST Antibody (Rabbit) (store at -20°C)
• LXR alpha-LBD, GST (-80°C)
• Fluoroscein Coactivator Peptide (-20°C)
• TR-FRET Coregulatore Buffer H (4°C)
• 1M DTT (-20°C or -80°C)
Assay Category Biochemical Assay
Druggable Target Nuclear Receptors
Readout TR-FRET (Activity)
Product Line LanthaScreen
For Use With (Application) Nuclear Receptor Assay, TR-FRET
Ligand LXR alpha
Product Type TR-FRET LXR Alpha Coactivator Assay Kit
No. of Assays 800 x 20 μL Assays
Mostrar más Mostrar menos

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.